missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford POPSO
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 68189-43-5
Molecular Formula: C10H22N2O8S2
Molecular Weight: 362.4Specifications
Specifications
| Chemical Name or Material | POPSO |
| CAS | 68189-43-5 |
| Quantity | 100 g |
| Molecular Formula | C10H22N2O8S2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction