Learn More
Cytiva Polyvinyl Chloride Fume Cupboard
This rich, robust, reliable, fume cupboard made of polyvinylchloride features glare fluorescent tube lightening.
Brand:  Cytiva 2649
Additional Details : Weight : 220.00000kg
 View more versions of this product
Product Code. 12506606
  • 6230.00€ / Each
Estimated Shipment: 24-05-2024
Description
  • Fume cupboard made of polyvinylchloride with glare fluorescent tube lightening
  • Standard sorbridge PVC (= registry fume cupboards)
  • PVC rear panel over the entire width of the fume hood
  • Guide the air at the top and bottom
  • Uses all types of solvents, volatile products, powders, condensing
Specifications
Specifications
Fume Cupboard
PVC
1 Ea.
Glare Fluorescent Tube
NFX 15-210 class B & EN 14175
180 cm