Learn More
Buerkle™ Polypropylene Sterilizable Bowl
Shop All Bürkle Products
Click to view available options
Diameter (Metric):
6 mm
Unit Size:
Each
Description
- Stackable
- Solidified substances are easily removed
- Polypropylene, white
- Autoclavable up to +121°C for 20min.
- Safe for foodstuff
Specifications
Specifications
| Description | Hose Nozzle |
| Color | Blue |
| Material | Polypropylene |
| Diameter (Metric) | 6 mm |