Learn More
Equip Labo™ Polypropylene Laboratory Fume Hood
Product Code. p-8009239
Click to view available options
Width (Metric):
120 cm
150 cm
180 cm
Unit Size:
Each
Product Code. 12576606
Brand: Equip Labo™ 2654
Rich, robust, reliable laboratory fume hood made of polypropylene material.
  • Melamine walls on standard fume cupboards and white water-repellent laminate with fire rating M1 on laboratory and attack fume cupboards 
  • Sécurit ™ glass folding window thickness 8mm 
  • Locking of the opening to 40 cm from the work surface 
  • Fluorescent tube lighting without glare 
  • Control panel, anemometer and audible and visual alarm on the right post of the fume cupboard 
  • Connection to extraction 200mm diameter 
  • Front average speed 0,3 m / s minimum 
  • Polypropylene back panel more chemically resistant 
Specifications
Product Type Laboratory Fume Hood
Lighting Type Glare Fluorescent Tube
Material Polypropylene
Certifications/Compliance XPX 15-206
Quantity 1 Ea.
Width (Metric) 120 cm