Learn More
Equip Labo™ Polypropylene Laboratory Fume Hood
Product Code. 12596606
Change view
Click to view available options
Width (Metric):
120 cm
150 cm
180 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
12596606 180 cm Each
12576606 120 cm Each
12556186 150 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12596606 Supplier Equip Labo™ Supplier No. 2656
  • ONLINE EXCLUSIVE PRICE
    6500.00€
    5438.05€ / Pack of 1
    Save 1061.95€
    16% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Rich, robust, reliable laboratory fume hood made of polypropylene material.
  • 180 x 80 x 155 cm dimensions
  • Back panel in polypropylene more chemically resistant
  • Gutter for the recovery of condensates to be connected to the grid
  • discharge
  • Fume hood made of polypropylene material
Specifications
Product Type Laboratory Fume Hood
Lighting Type Glare Fluorescent Tube
Material Polypropylene
Certifications/Compliance XPX 15-206
Quantity 1 Ea.
Width (Metric) 180 cm