Learn More
Equip Labo™ Polypropylene Laboratory Fume Hood
Click to view available options
Width (Metric):
120 cm
150 cm
180 cm
Unit Size:
Each
Description
- 180 x 80 x 155 cm dimensions
- Back panel in polypropylene more chemically resistant
- Gutter for the recovery of condensates to be connected to the grid
- discharge
- Fume hood made of polypropylene material
Specifications
Specifications
| Product Type | Laboratory Fume Hood |
| Lighting Type | Glare Fluorescent Tube |
| Material | Polypropylene |
| Certifications/Compliance | NFX 15-210 class B & EN 14175 |
| Quantity | 1 Ea. |
| Width (Metric) | 150 cm |