Learn More
Vitlab™ Polypropylene Bowl
Product Code. 10528341
Click to view available options
Capacity (Metric):
1 L
2 L
4 L
7 L
Unit Size:
Each
Pack of 3
Pack of 5
Product Code. 10528341
Brand: Vitlab™ 42694
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these bowls to help prevent splashing on the work surface while working with strong reactions in uncovered vessels. VITLAB™ Polypropylene Bowls feature a broad, stable edge and circumferential standing ring on the bottom.
  • White round bowl
  • Polypropylene
Specifications
Bottom Shape Flat
Capacity (Metric) 2 L
Color White
Material Polypropylene
Shape Round
Type Bowl
Height (Metric) 80 mm
Diameter (Metric) 200 mm