Learn More
Integra Biosciences™ Polypropylene, Bottle and Lid with PVDF Quick Couplings and Level Detection
Product Code. 10532043
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10532043 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10532043 Supplier Integra Biosciences™ Supplier No. 158431
  • ONLINE EXCLUSIVE PRICE
    675.00€
    621.78€ / Pack of 1
    Save 53.22€
    7% Off
Estimated Shipment: 10-12-2026
Capacity Metric: 4 L
Specifications
Capacity (Metric) 4 L
Coating Polypropylene
Material Polypropylene
Product Type Reservoir Bottle