Learn More
Thermo Scientific Chemicals Polyethylene Glove Bag, 68cm (27in) x 43cm (17in) x 30cm (12in), 56cm (22in) entrance
Product Code. 15474135
Change view
Click to view available options
Dimensions (L x W x H):
17 x 17 x 11 in.
27 x 17 x 12 in.
27 x 27 x 15 in.
37 x 27 x 18 in.
37 x 37 x 25 in.
Quantity:
1 Ea.
4 Pk.
Unit Size:
Each
Pack of 4
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Quantity Unit Size
15474135 37 x 27 x 18 in. 4 Pk. Pack of 4
11443880 17 x 17 x 11 in. 1 Ea. Each
11463880 27 x 17 x 12 in. 1 Ea. Each
11473880 27 x 17 x 12 in. 4 Pk. Pack of 4
11440960 27 x 27 x 15 in. 1 Ea. Each
11483880 27 x 27 x 15 in. 4 Pk. Pack of 4
15464135 37 x 27 x 18 in. 1 Ea. Each
11413890 37 x 37 x 25 in. 1 Ea. Each
11403890 37 x 37 x 25 in. 4 Pk. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15474135 Supplier Thermo Scientific Chemicals Supplier No. 093727.LL
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
For use with handling air/moisture sensitive high purity chemicals or materials. Ideal for laboratory use and field sampling.
Polyethylene Glove Bag is used for handling the air and moisture sensitive chemicals in laboratories. It is also used for field sampling and minimizes work exposure. Further, it preserves the integrity of the sample during storage. In addition to this, it serves as a portable laboratory for conducting tests and crime scene investigations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • A portable alternative to standard glove boxes
  • Includes closure, tubing coupler, and cotton gloves
  • Ideal for handling air/moisture sensitive, high purity chemicals or materials
  • For use in the laboratory and/or field sampling
  • Available in a variety of sizes and quantities
TRUSTED_SUSTAINABILITY
Specifications
Material Polyethylene
Color Clear
Dimensions (L x W x H) 37 x 27 x 18 in.
For Use With (Application) Field Sampling, Laboratory Use
Includes Closure, tubing coupler, and cotton gloves
Length (English) 37 in.
Quantity 4 Pk.