missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3F Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76929-0.025mg
This item is not returnable.
View return policy
Description
POLR3F Polyclonal specifically detects POLR3F in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| POLR3F | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DNA-directed RNA polymerase III 39 kDa polypeptide, DNA-directed RNA polymerase III subunit F, DNA-directed RNA polymerase III subunit RPC6, DNA-directed RNA polymerases III 39 kDa polypeptide, polymerase (RNA) III (DNA directed) polypeptide F (39 kDa), polymerase (RNA) III (DNA directed) polypeptide F, 39 kDa, RNA polymerase III 39 kDa subunit, RNA polymerase III C39 subunit, RNA polymerase III subunit C6, RPC39MGC13517, RPC6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10621 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| CAC11110 | |
| POLR3F | |
| Antibody was raised against a 21 amino acid synthetic peptide from near the amino terminus of human POLR3F. The immunogen is located within the first 50 amino acids of POLR3F. Amino Acid Squence: MAEVKVKVQPPDADPVEIEN | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction