missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2G Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47329-25ul
This item is not returnable.
View return policy
Description
POLR2G Polyclonal specifically detects POLR2G in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| POLR2G | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| DNA directed RNA polymerase II 19 kda polypeptide, DNA-directed RNA polymerase II 19 kDa polypeptide, DNA-directed RNA polymerase II subunit G, DNA-directed RNA polymerase II subunit RPB7, hRPB19, hsRPB7, MGC138369, polymerase (RNA) II (DNA directed) polypeptide G, RNA polymerase II 19 kDa subunit, RNA polymerase II seventh subunit, RNA polymerase II subunit B7, RPB19, RPB7MGC138367 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5436 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| POLR2G | |
| This antibody was developed against a recombinant protein corresponding to amino acids: NLLNTVKQKLFTEVEGTCTGKYGFVIAVTTIDNIGAGVIQPGRGFVLYPVKYKAIVFRPFKGEVVDAVVTQVNKVGLFTEIGPMSCFISRHSIPSEMEFDPNSNPPCYKTMDEDIVIQQDDEIRLKIVGTRVDKNDIFAIGSLMD | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction