missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99015-50ul
This item is not returnable.
View return policy
Description
POLG2 Polyclonal antibody specifically detects POLG2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| POLG2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| DNA polymerase gamma accessory 55 kDa subunit, DNA polymerase subunit gamma-2, mitochondrial, EC 2.7.7.7, HP55, Mitochondrial DNA polymerase accessory subunit, mitochondrial DNA polymerase subunit gamma-2, mitochondrial DNA polymerase, accessory subunit, MtPolB, MTPOLBPOLG-BETA, p55, PEOA4, POLB, POLGB, PolG-beta, polymerase (DNA directed), gamma 2, accessory subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human POLG2. | |
| 50 μg | |
| Endocrinology | |
| 11232 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction