Learn More
Podocan Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93238-0.02ml
This item is not returnable.
View return policy
Description
Podocan Polyclonal antibody specifically detects Podocan in Human, Mouse samples. It is validated for Western Blot
Specifications
| Podocan | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| PCAN, PODOCAN, podocan proteoglycan, SLRR5AMGC24995 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 67-210 of human PODN (NP_714914.2). LGPVLAVRAPGFGRSGGHSLSPEENEFAEEEPVLVLSPEEPGPGPAAVSCPRDCACSQEGVVDCGGIDLREFPGDLPEHTNHLSLQNNQLEKIYPEELSRLHRLETLNLQNNRLTSRGLPEKAFEHLTNLNYLYLANNKLTLAP | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 127435 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |