Learn More
PNMT Antibody (CL11434) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21193MFV610
Product Code. 30297724
  • 780.00€ / 0.10mL
Description
PNMT Monoclonal antibody specifically detects PNMT in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PNMT
Monoclonal
mFluor Violet 610 SE
EC 2.1.1, EC 2.1.1.28, Noradrenaline N-methyltransferase, PENTMGC34570, phenylethanolamine N-methylase, phenylethanolamine N-methyltransferase, PNMTase
This antibody was developed using a recombinant protein derived from P11086, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
5409
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL11434
50mM Sodium Borate
Mouse
Protein A purified