missing translation for 'onlineSavingsMsg'
Learn More

PNMA3 Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene

Product Code. 17576961
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17576961 30 μL 30µL
17556961 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17576961 Supplier Origene Technologies Supplier No. TA502466S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PNMA3 Monoclonal antibody specifically detects PNMA3 in Human samples. It is validated for Western Blot

Specifications

Antigen PNMA3
Applications Western Blot
Classification Monoclonal
Clone OTI2D3
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene BTRC
Gene Accession No. Q9Y297
Gene Alias MA3; MA5; Paraneoplastic antigen Ma3; paraneoplastic antigen Ma3 homolog; paraneoplastic cancer-testis-brain antigen; paraneoplastic Ma antigen 3; PNMA3; RGD1563752
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 52-354 of human BTRC produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8945
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.