missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNK Antibody (SR2021), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22588-100ul
This item is not returnable.
View return policy
Description
PNK Monoclonal antibody specifically detects PNK in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| PNK | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR2021 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| DNA 5'-kinase/3'-phosphatase, EIEE10, Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP), MCSZ, PNKbifunctional polynucleotide phosphatase/kinase, polynucleotide kinase 3'-phosphatase, Polynucleotide kinase-3'-phosphatase | |
| A synthesized peptide derived from human PNK (Uniprot #: Q96T60) | |
| 100 μg | |
| DNA Repair, Protein Phosphatase, Signal Transduction | |
| 11284 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction