missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMEL17/SILV Mouse anti-Human, Clone: PMEL/2037, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | PMEL17/SILV |
|---|---|
| Clone | PMEL/2037 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 2-4 μg/mL, ELISA 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Description
PMEL17/SILV Monoclonal specifically detects PMEL17/SILV in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
| PMEL17/SILV | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
| A recombinant fragment (around aa 376-502) of human PMEL17/SILV protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| PMEL/2037 | |
| Western Blot 2-4 μg/mL, ELISA 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6490 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title