missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMCA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
200.00€
Specifications
| Antigen | PMCA2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
PMCA2 Polyclonal specifically detects PMCA2 in Human, Rat, Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| PMCA2 | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q01814, P11506 | |
| 491 | |
| Synthetic Peptide: T(5) N S D F Y S K N Q R N E S S(19) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
| plasma membrane calcium-transporting ATPase 2, ATPase, Ca++ transporting, plasma membrane 2, PMCA2, PMCA2a, PMCA2i | |
| ATP2B2 | |
| IgG | |
| Affinity Purified | |
| PMCA2 antibody detects calcium pump of the plasma membrane 2 (PMCA2 ATPase) from human and rat and porcine tissues and cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title