Learn More
Invitrogen™ PLXNA4 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149079
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
PLXNA4 is a protein coding gene. Coreceptor for SEMA3A. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
Specifications
| PLXNA4 | |
| Polyclonal | |
| Unconjugated | |
| Plxna4 | |
| 9330117B14; FAYV2820; Kiaa1550; mKIAA1550; OTTHUMP00000207245; PLEXA4; plexin A4; plexin A4, A; plexin-A4; Plxa4; Plxna4; Plxna4a; PLXNA4B; PRO34003; UNQ2820/PRO34003 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 91584 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9HCM2 | |
| Plxna4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Plexin A4, Thr24-Pro1237. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |