missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLTP Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148961
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
PLTP is one of at least two lipid transfer proteins found in human plasma. The protein transfers phospholipids from triglyceride-rich lipoproteins to high density lipoprotein (HDL). In addition to regulating the size of HDL particles, this protein may be involved in cholesterol metabolism.
Specifications
| PLTP | |
| Polyclonal | |
| Unconjugated | |
| PLTP | |
| BPI fold containing family E; BPIFE; HDLCQ9; lipid transfer protein II; OD107; Phospholipid transfer protein; Pltp | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 5360 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P55058 | |
| PLTP | |
| Mouse myeloma cell line NS0-derived recombinant human PLTP isoform 1, Glu18-Val493 (Glu18Val). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?