missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLTP Antibody (SR2240), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00€ - 449.00€
Specifications
| Antigen | PLTP |
|---|---|
| Clone | SR2240 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641185
|
Novus Biologicals
NBP3-22273-100ul |
100 μg |
449.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633144
|
Novus Biologicals
NBP3-22273-50ul |
50 μg |
338.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLTP Monoclonal antibody specifically detects PLTP in Human, Rat samples. It is validated for Western BlotSpecifications
| PLTP | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| HDLCQ9, Lipid transfer protein II, phospholipid transfer protein | |
| A synthesized peptide derived from human PLTP (Uniprot #: P55058) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2240 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5360 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title