missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55088
This item is not returnable.
View return policy
Description
PLK4 Polyclonal specifically detects PLK4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| PLK4 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| EC 2.7.11.21, PLK-4, polo-like kinase 4 (Drosophila), polo-like kinase 4SAKSTK18serine/threonine kinase 18, Sak, serine/threonine protein kinase SAK, Serine/threonine-protein kinase 18, serine/threonine-protein kinase PLK4, Serine/threonine-protein kinase Sak, Snk akin kinase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| PLK4 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RPPFDTDTVKNTLNKVVLADYEMPSFLSIEAKDLIHQLLRRNPADRLSLSSVLDHPFMSRNSSTKSKDLGTVEDSIDSGHATISTAITASSSTSISGSLFDKRRLLIGQPLPNKMTVFPKNKSSTDFSSSGDGNS | |
| 100 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
| 10733 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction