missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 Antibody (AZ44) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00€ - 453.00€
Specifications
| Antigen | PLK1 |
|---|---|
| Clone | AZ44 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18542564
|
Novus Biologicals
NB100-2747-0.025mg |
0.025 mg |
190.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18541832
|
Novus Biologicals
NB100-2747 |
0.1 mg |
453.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLK1 Monoclonal antibody specifically detects PLK1 in Xenopus samples. It is validated for Western Blot, ELISASpecifications
| PLK1 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
| PBS | |
| 5347 | |
| IgG1 κ | |
| Protein A purified |
| AZ44 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Xenopus species | |
| cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
| Baculo-expressed PLK1 from Xenopus laevis | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title