missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 Antibody (13E8), Novus Biologicals™
Mouse Monoclonal Antibody
213.00€
Specifications
| Antigen | PLK1 |
|---|---|
| Clone | 1300000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 |
| Applications | Western Blot |
Description
PLK1 Monoclonal specifically detects PLK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| PLK1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
| PLK1 | |
| IgG2b | |
| Protein G purified | |
| Detects PLK1 from human, mouse, and rat samples. |
| 1300000000 | |
| Western Blot 1:1000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P53350 | |
| 5347 | |
| Recombinant protein containing residues 300-400 of human PLK1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title