missing translation for 'onlineSavingsMsg'
Learn More

PlGF-3 Antibody (1039018) [HRP], Novus Biologicals™

Product Code. 30644800 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30644800 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30644800 Supplier Novus Biologicals Supplier No. FAB7758H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PlGF-3 Monoclonal antibody specifically detects PlGF-3 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PlGF-3
Applications Immunohistochemistry
Classification Monoclonal
Clone 1039018
Conjugate HRP
Formulation PBS
Host Species Mouse
Immunogen Spodoptera frugiperda Sf21 (baculovirus)-derived human PlGF-3, Leu19-Arg221, Accession # P49763
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5281
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.