missing translation for 'onlineSavingsMsg'
Learn More

PLC-delta 3 Antibody, Novus Biologicals™

Product Code. 18691809 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18691809 25 μL 25µL
18608008 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18691809 Supplier Novus Biologicals Supplier No. NBP26870625ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLC-delta 3 Polyclonal antibody specifically detects PLC-delta 3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLC-delta 3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias EC 3.1.4.11, KIAA1964, MGC71172, Phosphoinositide phospholipase C-delta-3, phospholipase C, delta 3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3, Phospholipase C-delta-3, PLC delta3, PLC-delta-3
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: EGHQSEGLRRFGGAFAPARCLTIAFKGRRKNLDLAAPTAEEAQRWVRGLTKLRARLDAMSQRERLDHWIHSYLHRADSNQDSKMSFKEIKSLLRMVNVDMNDMYAYLLFK
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 113026
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.