missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plakophilin 1 Antibody (10B2), Novus Biologicals™
Mouse Monoclonal Antibody
265.00€ - 518.00€
Specifications
| Antigen | Plakophilin 1 |
|---|---|
| Clone | 10B2 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18215020
|
Novus Biologicals
NBP2-53326-20UG |
20 μg |
265.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18253610
|
Novus Biologicals
NBP2-53326-100UG |
100 μg |
518.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plakophilin 1 Monoclonal specifically detects Plakophilin 1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| Plakophilin 1 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| Unconjugated | |
| Mouse | |
| Cellular Markers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 5317 | |
| N-terminus of human Plakophilin-1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| 10B2 | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| B6PBand 6 protein, MGC138829, plakophilin 1 (ectodermal dysplasia/skin fragility syndrome), plakophilin-1 | |
| PKP1 | |
| IgG1 | |
| Protein A or G purified | |
| 75 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title