missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2R1 Antibody (CL0485) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52934MFV450
This item is not returnable.
View return policy
Description
PLA2R1 Monoclonal antibody specifically detects PLA2R1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PLA2R1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 180 kDa secretory phospholipase A2 receptor, CLEC13CC-type lectin domain family 13 member C, phospholipase A2 receptor 1, 180kDa, PLA2G1R, PLA2IR, PLA2-RM-type receptor, PLA2Rphospholipase A2 receptor 1, 180kD, secretory phospholipase A2 receptor | |
| This antibody was developed using a recombinant protein derived from Q13018, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0485 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 22925 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction