missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G1B Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90537-100UL
This item is not returnable.
View return policy
Description
PLA2G1B Monoclonal specifically detects PLA2G1B in Mouse samples. It is validated for ELISA, Sandwich ELISA Capture.
Specifications
| PLA2G1B | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 1 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 | |
| Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835 | |
| Recombinant Mouse PLA2G1B protein | |
| 100 μg | |
| Lipid and Metabolism | |
| 5319 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction