missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKR Antibody (HL71/10) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1980W
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PKR Monoclonal antibody specifically detects PKR in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry
Spezifikation
| PKR | |
| Monoclonal | |
| DyLight 650 | |
| double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated | |
| Protein kinase fraction from interferon-treated Daudi human Burkitt's lymphoma cell line. Laurent, A.G. et al. (1985) Proc. Natl. Acad. Sci. 82:4341., | |
| 0.1 mL | |
| Apoptosis, Phospho Specific, Signal Transduction | |
| 5610 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| HL71/10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur