missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKM2 Antibody (SR2031), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00€ - 449.00€
Specifications
| Antigen | PKM2 |
|---|---|
| Clone | SR2031 |
| Dilution | Western Blot 1:1000-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641475
|
Novus Biologicals
NBP3-21688-100ul |
100 μg |
449.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651975
|
Novus Biologicals
NBP3-21688-50ul |
50 μg |
338.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKM2 Monoclonal antibody specifically detects PKM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| PKM2 | |
| Western Blot 1:1000-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, PK, muscle type, PK2, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK | |
| A synthesized peptide derived from human PKM2 (Uniprot #: P14618) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2031 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5315 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title