Learn More
Novus Biologicals™ PKC beta 2 Antibody (SR2824), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27716-50ul
This item is not returnable.
View return policy
Description
PKC beta 2 Monoclonal antibody specifically detects PKC beta 2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| PKC beta 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR2824 | |
| Western Blot 1:500-1:2000, Flow Cytometry, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| PKCB;PKCbeta;PKC-beta;PKCI(2);PRKCB;PRKCB1;PRKCB2;Protein kinase C beta type | |
| A synthesized peptide derived from human PKC beta 2 (Uniprot #: P05771) | |
| 50 μL | |
| Cancer, mTOR Pathway, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 5579 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |