Learn More
Pirlimicyn hydrochloride 1000 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Pirlimycin Hydrochloride |
| CAS | 78822-40-9 |
| Analyte or Component Names | Pirlimycin Hydrochloride |
| Shipping Condition | +20°C |
| Molecular Formula | C17 H31 Cl N2 O5 S . Cl H |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[(4-ethyl-2-piperidinyl)carbonyl]amino]-1-thio-, monohydrochloride, (2S-cis)-, L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-4-ethyl-2-piperidinyl]carbonyl]amino]-1-thio-, monohydrochloride (9CI), Pirlimycin hydrochloride, Pirsue, U 57930E |
| InChI Formula | InChI=1S/C17H31ClN2O5S.ClH/c1-4-9-5-6-19-10(7-9)16(24)20-11(8(2)18)15-13(22)12(21)14(23)17(25-15)26-3;/h8-15,17,19,21-23H,4-7H2,1-3H3,(H,20,24);1H/t8-,9+,10-,11+,12-,13+,14+,15+,17+;/m0./s1 |
| SMILES | Cl.CC[C@@H]1CCN[C@@H](C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O |
| Show More |