missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pirbuterol Dihydrochloride, Mikromol™
Description
Pirbuterol Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Pirbuterol Dihydrochloride |
| CAS | 38029-10-6 |
| Analyte or Component Names | Pirbuterol Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H20 N2 O3 . 2 Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Broncocor, CP 24314-1, Exirel, Exirel (bronchodilator), NSC 355078, Pirbuterol dihydrochloride, Pirbuterol hydrochloride, Pyrbuterol hydrochloride, 2,6-Pyridinedimethanol, alpha6-[[(1,1-dimethylethyl)amino]methyl]-3-hydroxy-, hydrochloride (1:2), 2,6-Pyridinedimethanol, alpha6-[[(1,1-dimethylethyl)amino]methyl]-3-hydroxy-, dihydrochloride (9CI) |
| InChI Formula | InChI=1S/C12H20N2O3.2ClH/c1-12(2,3)13-6-11(17)8-4-5-10(16)9(7-15)14-8;;/h4-5,11,13,15-17H,6-7H2,1-3H3;2*1H |
| SMILES | Cl.Cl.CC(C)(C)NCC(O)c1ccc(O)c(CO)n1 |
| IUPAC Name | 6-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)pyridin-3-ol;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?