Learn More
Eppendorf™ Pipette Lower Part For Adjustable Pipette
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
0.5 to 10 μL 12-channel Pipette
10 mL Single-channel Pipette
1200 μL Multichannel Pipette
15 to 300 μL 8-channel Pipette
5 mL Single-channel Pipette
5 to 100 μL 12-channel Pipette
5 to 100μL 8-channel Adjustable Pipette
For 0.5 to 10μL 8-channel Adjustable Pipette
Unit Size:
Each
Description
- Accessory for pipettes
- Lower part for adjustable pipettes
- Available in a wide range of sizes to suit a variety of applications
Specifications
Specifications
| Color | Orange |
| Type | Pipette Lower Part |
| For Use With (Equipment) | 15 to 300 μL 8-channel Pipette |
| Quantity | 1 Each |