Learn More
Eppendorf™ Pipette Lower Part For Adjustable Pipette
Product Code. 10230370 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
0.5 to 10 μL 12-channel Pipette
10 mL Single-channel Pipette
1200 μL Multichannel Pipette
15 to 300 μL 8-channel Pipette
5 mL Single-channel Pipette
5 to 100 μL 12-channel Pipette
5 to 100μL 8-channel Adjustable Pipette
For 0.5 to 10μL 8-channel Adjustable Pipette
Unit Size:
Each
Product Code. 10230370
Brand: Eppendorf™ 4861608005
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for use with Eppendorf Pipettes
  • Accessory for pipettes
  • Lower part for adjustable pipettes
  • Available in a wide range of sizes to suit a variety of applications
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Type Pipette Lower Part
For Use With (Equipment) 5 to 100μL 8-channel Adjustable Pipette
Quantity 1 Each