Description
- Problem-free pipetting of problem liquids.
- Specially suited for liquids with high densities, viscosities or vapor pressures.
- Given the disposable capillaries and pistons, positive displacement absolutely eliminates cross-contamination from aerosols.
- Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
Specifications
Specifications
| Volume (Metric) | 10 to 100 μL |
| Volume (Metric) Dispensing | 10/100 μL |
| For Use With (Application) | Specially suited for liquids with high densities, viscosities, or vapor pressures. |
| Quantity | 1/Pk |
| Product Type | Pipette |