missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PIPES, 0.5M buffer soln., pH 6.5

Product Code. p-7042058
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483839 100 mL 100mL
15493839 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483839 Supplier Thermo Scientific Chemicals Supplier No. J61224.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

PIPES can be used in a wide array of biological applications, including as a component in media formulations. Specific applications include buffers for cell culture and diagnostic reagent production. It is used to buffer glutaraldehyde fixative solutions when fixing plant and/or animal tissue tissue samples. This will help prevent the loss of lipids. It is used to buffer glutaraldehyde and formaldehyde when fixing fungal zoospores for fluorescence microscopy and electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES can be used in a wide array of biological applications, including as a component in media formulations. Specific applications include buffers for cell culture and diagnostic reagent production. It is used to buffer glutaraldehyde fixative solutions when fixing plant and/or animal tissue tissue samples. This will help prevent the loss of lipids. It is used to buffer glutaraldehyde and formaldehyde when fixing fungal zoospores for fluorescence microscopy and electron microscopy.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents, bases and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
MDL Number MFCD00006159
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Color Colorless
Concentration 0.5 M
pH 6.5
Physical Form Liquid
Quantity 100 mL
Solubility Information Fully miscible in water.
Chemical Name or Material PIPES buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.