missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP5K2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38497-20ul
This item is not returnable.
View return policy
Description
PIP5K2B Polyclonal antibody specifically detects PIP5K2B in Human samples. It is validated for ELISA,Western Blot,Immunofluorescence
Specifications
| PIP5K2B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunofluorescence 1:50 - 1:200 | |
| 1-phosphatidylinositol-4-phosphate kinase, 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta, Diphosphoinositide kinase 2-beta, EC 2.7.1, EC 2.7.1.149, phosphatidylinositol-4-phosphate 5-kinase, type II, beta, Phosphatidylinositol-5-phosphate 4-kinase type II beta, phosphatidylinositol-5-phosphate 4-kinase type-2 beta, phosphatidylinositol-5-phosphate 4-kinase, type II, beta, PI(5)P 4-kinase type II beta, PI5P4KB, PIP4KII-beta, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PTDINS(4)P-5-kinase, PtdIns(5)P-4-kinase isoform 2-beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human PIP5K2B (NP_003550.1).,, Sequence:, MSSNCTSTTAVAVAPLSASKTKTKKKHFVCQKVKLFRASEPILSVLMWGVNHTINELSNVPVPVMLMPDDFKAYSKIKVDNHLFNKENLPSRFKFKEYCPMVFRNLRERFGIDDQDYQNSVTRSAPINSDSQGRCGTRFLTTYDRRFVIKTVSSEDVAEMHNILKKYHQFIVECHGNTLLPQFLGMYRLTVDGVETYMVVTRNVFSHRLTVHRKYDLKGSTVAREASDKEKAKDLPTFKDNDFLNEGQKLHVGEESKKNFLEKLKRDVEF | |
| 20 μL | |
| Cancer | |
| 8396 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction