missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP5K1C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | PIP5K1C |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16845755
|
Proteintech
27640-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16835755
|
Proteintech
27640-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphatidylinositol-4-phosphate-5-kinase (PIPK) synthesizes phosphatidylinositol-4,5-bisphosphate, which regulates various processes including cell proliferation, survival, membrane trafficking and cytoskeletal organization. The PIPK family is divided into type I, type II and type III. Each type of the PIPK family phosphorylates distinct substrates. They contain an activation loop, which determines their enzymatic specificity and subcellular targeting. The phosphatidylinositol-4-phosphate-5-kinase type I consists of three members, PIPK I alpha, beta and gamma, which are characterized by phosphorylating PI4P on the 5-hydroxyl. PIPK I alpha, designated PIPK I beta in mouse, is expressed in brain tissue. PIPK I beta, designated PIPK I alpha in mouse, is also called STM7. PIPK I gamma has two variants produced by alternative splicing which are expressed in lung, brain and kidneys.Specifications
| PIP5K1C | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O60331 | |
| 23396 | |
| PIP5K1C Fusion Protein Ag26525 | |
| Primary | |
| -20°C | |
| PIP5K1C |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| KIAA0589, LCCS3, PIP5K GAMMA, PIP5K1 gamma, PIP5K1C, PIP5Kgamma, PIP5KIgamma, PtdIns(4)P 5 kinase 1 gamma | |
| PIP5K1C | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title