missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PINK1 Protein

Product Code. 18556151 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
This item is not returnable. View return policy

Product Code. 18556151

Brand: Novus Biologicals™ NBP1724810.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

An un-tagged denatured recombinant protein corresponding to the amino acids 156-507 of Human PINK1 The Recombinant Human PINK1 Protein is derived from E. coli. The Recombinant Human PINK1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_115785
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) 1M Urea, 5% Glycerol.
Gene ID (Entrez) 65018
Molecular Weight (g/mol) 37.9kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Recombinant human PINK protein was expressed in E. coli
Immunogen PINK1, 156-507 aa, Human, E. coli (NP_115785).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PINK1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.