missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TSC22D4 is a leucine zipper-containing protein that is highly conserved during evolution. It is transcriptionally up-regulated by many different stimuli, including anti-cancer drugs and growth inhibitors. TSC22D4 may play a suppressive role in tumorigenesis.
Specifications
Specifications
| Antigen | Pinin |
| Applications | Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PNN |
| Gene Accession No. | Q9H307 |
| Gene Alias | Desmosome associated protein, Domain rich serine protein, DRS, DRS protein, DRSP, memA, Nuclear protein SDK3, Pinin, PNN, SDK3, SR like protein |
| Gene Symbols | PNN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?