Learn More
Pinic Acid (Diastereomeric Mixture), TRC
Product Code. 30135652
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135652 500 mg 500mg
30005342 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135652 Supplier Toronto Research Chemicals Supplier No. TRCP468200500MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28664-02-0
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
Synonym 3-Carboxy-2,2-dimethyl-cyclobutaneacetic acid, NSC 2802
IUPAC Name 3-(carboxymethyl)-2,2-dimethylcyclobutane-1-carboxylic acid
SMILES O=C(O)C1CC(CC(O)=O)C1(C)C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C9H14O4/c1-9(2)5(4-7(10)11)3-6(9)8(12)13/h5-6H,3-4H2,1-2H3,(H,10,11)(H,12,13)
Formula Weight 186.0892
Percent Purity >95%
Chemical Name or Material Pinic Acid (Diastereomeric Mixture)
Grade HPLC