CAS RN 28664-02-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pinic Acid (Diastereomeric Mixture), TRC
CAS: 28664-02-0 Molecular Formula: C9H14O4 Molecular Weight (g/mol): 186.21 Synonym: 3-Carboxy-2,2-dimethyl-cyclobutaneacetic acid,NSC 2802 IUPAC Name: 3-(carboxymethyl)-2,2-dimethylcyclobutane-1-carboxylic acid SMILES: O=C(O)C1CC(CC(O)=O)C1(C)C