missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIN/DLC8 Antibody (20A1), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | PIN/DLC8 |
|---|---|
| Clone | 20A1 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043997
|
Novus Biologicals
NBP3-26610-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043722
|
Novus Biologicals
NBP3-26610-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIN/DLC8 Monoclonal antibody specifically detects PIN/DLC8 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
| PIN/DLC8 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cytoplasmic dynein light polypeptide, DLC1DNCLC1, DLC8MGC126137, DNCL1MGC126138, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, hdlc1, LC8, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase | |
| A synthesized peptide derived from Human PIN/DLC8 [UniProt P63167] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 20A1 | |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| p53 Pathway | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 8655 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title