missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PIK3R1 (Human) Recombinant Protein
Human PIK3R1 full-length ORF ( AAH30815, 1 a.a. - 454 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005295-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MYNTVWNMEDLDLEYAKTDINCGTDLMFYIEMDPPALPPKPPKPTTVANNGMNNNMSLQDAEWYWGDISREEVNEKLRDTADGTFLVRDASTKMHGDYTLTLRKGGNNKLIKIFHRDGKYGFSDPLTFSSVVELINHYRNESLAQYNPKLDVKLLYPVSKYQQDQVVKEDNIEAVGKKLHKYNTQFQEKSREYDRLYEEYTRTSQEIQMKRTAIEAFNETIKIFEEQCQTQERYSKEYIEKFKREGNEKEIQRIMHNYDKLKSRISEIIDSRRRLEEDLKKQAAEYREIDKRMNSIKPDLIQLRKTRDQYLMWLTQKGVRQKKLNEWLGNENTEDQYSLVEDDEDLPHHDEKTWNVGSSNRNKAENLLRGKRDGTFLVRESSKQGCYACSVVVDGEVKHCVINKTATGYGFAEPYNLYSSLKELVLHYQHTSLVQHNDSLNVTLAYPVYAQQRR
Specifications
AAH30815 | |
phosphoinositide-3-kinase, regulatory subunit 1 (alpha) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PIK3R1 | |
GST |
5295 | |
Wheat germ expression system | |
10 μg | |
GRB1, p85, p85-ALPHA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |