missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PIK3R1 (Human) Recombinant Protein
Human PIK3R1 full-length ORF ( AAH30815, 1 a.a. - 454 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH30815 |
---|---|
Gene ID (Entrez) | 5295 |
Name | phosphoinositide-3-kinase, regulatory subunit 1 (alpha) |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16159111
|
Abnova™
H00005295-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16169111
|
Abnova™
H00005295-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
- Sequence: MYNTVWNMEDLDLEYAKTDINCGTDLMFYIEMDPPALPPKPPKPTTVANNGMNNNMSLQDAEWYWGDISREEVNEKLRDTADGTFLVRDASTKMHGDYTLTLRKGGNNKLIKIFHRDGKYGFSDPLTFSSVVELINHYRNESLAQYNPKLDVKLLYPVSKYQQDQVVKEDNIEAVGKKLHKYNTQFQEKSREYDRLYEEYTRTSQEIQMKRTAIEAFNETIKIFEEQCQTQERYSKEYIEKFKREGNEKEIQRIMHNYDKLKSRISEIIDSRRRLEEDLKKQAAEYREIDKRMNSIKPDLIQLRKTRDQYLMWLTQKGVRQKKLNEWLGNENTEDQYSLVEDDEDLPHHDEKTWNVGSSNRNKAENLLRGKRDGTFLVRESSKQGCYACSVVVDGEVKHCVINKTATGYGFAEPYNLYSSLKELVLHYQHTSLVQHNDSLNVTLAYPVYAQQRR
Specifications
AAH30815 | |
phosphoinositide-3-kinase, regulatory subunit 1 (alpha) | |
125% SDS-PAGE Stained with Coomassie Blue | |
GRB1, p85, p85-ALPHA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5295 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PIK3R1 | |
GST |