missing translation for 'onlineSavingsMsg'
Learn More

PIK3CG Monoclonal Antibody (OTI4B6), TrueMAB™, OriGene

Product Code. 17563402
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17563402 30 μL 30µL
17543402 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17563402 Supplier Origene Technologies Supplier No. TA505219S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIK3CG Monoclonal antibody specifically detects PIK3CG in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PIK3CG
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MRPS34
Gene Accession No. P82930
Gene Alias 1-phosphatidylinositol 3-kinase; 5830428L06Rik; EGK_14026; p110gamma; p110-gamma; p120-PI3K; phosphatidylinositol 3 kinase gamma, p110 gamma; phosphatidylinositol 3-kinase catalytic 110-kD gamma; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit gamma isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit gamma; Phosphoinositide-3-kinase catalytic gamma polypeptide; phosphoinositide-3-kinase gamma catalytic subunit; phosphoinositide-3-kinase, catalytic, gamma polypeptide; PI3CG; PI3K; pi3kcg; Pi3kg1; PI3Kgamma; PI3K-gamma; PI3-kinase subunit gamma; PIK3; Pik3cg; ptdIns-3-kinase subunit gamma; ptdIns-3-kinase subunit p110-gamma; Serine/threonine protein kinase PIK3CG
Host Species Mouse
Immunogen Full length human recombinant protein of human MRPS34 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65993
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.