missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGQ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59738
This item is not returnable.
View return policy
Description
PIGQ Polyclonal specifically detects PIGQ in Human samples. It is validated for Western Blot.
Specifications
| PIGQ | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| c407A10.1, c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)), class Q, EC 2.4.1.198, hGPI1, MGC12693, N-acetylglucosaminyl transferase component Gpi1, N-acetylglucosamyl transferase component GPI1, phosphatidylinositol glycan anchor biosynthesis, class Q, phosphatidylinositol N-acetylglucosaminyltransferase subunit Q, Phosphatidylinositol-glycan biosynthesis class Q protein, PIG-Q | |
| Rabbit | |
| 84 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BRB3 | |
| PIGQ | |
| Synthetic peptides corresponding to PIGQ(phosphatidylinositol glycan anchor biosynthesis, class Q) The peptide sequence was selected from the N terminal of PIGQ. Peptide sequence VLHFPFIPIQVKQLLAQVRQASQVGVAVLGTWCHCRQEPEESLGRFLESL. | |
| Affinity purified | |
| RUO | |
| 9091 | |
| Human, Mouse, Rat, Pig, Bovine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion