missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PIEZO1 Monoclonal antibody specifically detects PIEZO1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PIEZO1 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL9714 |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FAM38A, member A, membrane protein induced by beta-amyloid treatment, Mib, piezo-type mechanosensitive ion channel component 1 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from Q92508, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?